Learn More
ACROBiosystems ACROBiosystems Human HE4 / WFDC2 Protein, His Tag
Catalog No. 501956870
Encompass_Preferred
Recombinant Protein;100UG;Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Phe 124 (Accession # AAH46106).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 19 kDa and 22-26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-195-6870 Supplier ACROBiosystems Supplier No. HE4H5223100UG
Recombinant Protein;100UG;Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Phe 124 (Accession # AAH46106).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 19 kDa and 22-26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.