Learn More
Hoefer, Inc Hoefer™ HE99X Max Horizontal Agarose Electrophoresis Unit (Complete unit)
SDP
Catalog No. NC2740713
Encompass_Preferred
Offers a variety of gel lengths and comb sizes to enable the design of the gel required for optimum resolution of DNA fragmentsThe HE99X Max Agarose Electrophoresis Unit facilitates high resolution separation of nucleic acid fragments 100 bp to 20 kbp. Three different length trays are available for use with this unit.Features and BenefitsSelf-locating/centering guides—supports consistent running tray placementGel anchor feet—prevent the gel from floating or sliding during the runVariety of comb sizes—enables customization of sample throughput to fit your laboratory's requirementsAdjustable comb height—control well depthThree casting kits available—enabling users to run 15 x 20,15 x 15, or 15 x 10 cm gels in the same unitComplete Unit Includes:Buffer Chamber; Safety Lid with High Voltage Leads; Running Tray & Casting Tray, 15 x 20 cm; 15-well comb, 2 pcs; Comb Back, 2 pcs;Basic Unit Includes:The same as above without the Running Tray, Casting Tray, Comb, and Comb Back.
Catalog No. NC2740713 Supplier Hoefer, Inc Supplier No. HE99X151.5
Offers a variety of gel lengths and comb sizes to enable the design of the gel required for optimum resolution of DNA fragmentsThe HE99X Max Agarose Electrophoresis Unit facilitates high resolution separation of nucleic acid fragments 100 bp to 20 kbp. Three different length trays are available for use with this unit.Features and BenefitsSelf-locating/centering guides—supports consistent running tray placementGel anchor feet—prevent the gel from floating or sliding during the runVariety of comb sizes—enables customization of sample throughput to fit your laboratory's requirementsAdjustable comb height—control well depthThree casting kits available—enabling users to run 15 x 20,15 x 15, or 15 x 10 cm gels in the same unitComplete Unit Includes:Buffer Chamber; Safety Lid with High Voltage Leads; Running Tray & Casting Tray, 15 x 20 cm; 15-well comb, 2 pcs; Comb Back, 2 pcs;Basic Unit Includes:The same as above without the Running Tray, Casting Tray, Comb, and Comb Back.