Learn More
Ace Glass, Inc. Head, 100mm Duran flange, 5 neck, 29/42 center, (1)29/42, (3)24/40
SDP
Catalog No. 50978735
Encompass_Preferred
5 neck head with 100mm flat ground Duran flange. Head features one 29/42 center neck, three 24/40 side necks, and one 29/42 side neck.
Catalog No. 50-978-735 Supplier Ace Glass, Inc. Supplier No. 644308
5 neck head with 100mm flat ground Duran flange. Head features one 29/42 center neck, three 24/40 side necks, and one 29/42 side neck.