Learn More
Ace Glass, Inc. Head, 100mm Duran flange, 5 neck, 24/40 center, (3)24/29, (1)29/32
SDP
Catalog No. 50972992
Encompass_Preferred
5 neck head with 100mm flat ground Duran flange. Head features one 24/40 center neck, three 24/29 side necks, and one 29/32 side neck.
Catalog No. 50-972-992 Supplier Ace Glass, Inc. Supplier No. 6443108
5 neck head with 100mm flat ground Duran flange. Head features one 24/40 center neck, three 24/29 side necks, and one 29/32 side neck.