Learn More
Ace Glass, Inc. Head, 100mm conical flange, 4 neck, 45/50 center, (3)24/40
SDP
Catalog No. 50973106
Encompass_Preferred
4 neck reaction flask head with 4(100mm) Ace-Thred conical flange. 45/50 center neck in-line with two 24/40 side necks. Third 24/40 side neck is perpendicular to the in-line necks.
Catalog No. 50-973-106 Supplier Ace Glass, Inc. Supplier No. 649918
4 neck reaction flask head with 4(100mm) Ace-Thred conical flange. 45/50 center neck in-line with two 24/40 side necks. Third 24/40 side neck is perpendicular to the in-line necks.