Learn More
Ace Glass, Inc. Head, 100mm Duran flange, 5 neck, 45/50 center, (3)24/40, (1)29/42
SDP
Supplier:  Ace Glass, Inc. 644310
Encompass_Preferred
5 neck head with 100mm flat ground Duran flange. Head features one 45/50 center neck, three 24/40 side necks, and one 29/42 side neck.
Catalog No. 50-972-987