Learn More
Ace Glass, Inc. Head, 6in, four neck, 29/42 center, (3)24/40
SDP
Catalog No. 50973189
Encompass_Preferred
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has four necks: (1) 29/42 center, (3) 24/40 sides.
Catalog No. 50-973-189 Supplier Ace Glass, Inc. Supplier No. 652936
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has four necks: (1) 29/42 center, (3) 24/40 sides.