Learn More
Ace Glass, Inc. Head, 150mm Duran flange, 5 neck, 24/40 center, (3)24/40, (1)45/50
SDP
Catalog No. 50972988
Encompass_Preferred
5 neck head with 150mm flat ground Duran flange. Head features one 24/40 center neck, three 24/40 side necks, and one 45/50 side neck.
Catalog No. 50-972-988 Supplier Ace Glass, Inc. Supplier No. 644312
5 neck head with 150mm flat ground Duran flange. Head features one 24/40 center neck, three 24/40 side necks, and one 45/50 side neck.