Learn More
Ace Glass, Inc. Head, 150mm Duran flange, 5 neck, 45/50 center, (3)24/29, (1)45/40
SDP
Catalog No. 50978738
Encompass_Preferred
5 neck head with 150mm flat ground Duran flange. Head features one 45/50 center neck, three 24/29 side necks, and one 45/40 side neck.
Catalog No. 50-978-738 Supplier Ace Glass, Inc. Supplier No. 6443118
5 neck head with 150mm flat ground Duran flange. Head features one 45/50 center neck, three 24/29 side necks, and one 45/40 side neck.