Learn More
Ace Glass, Inc. Head, 6in, five neck, 24/40 center, (4)24/40
SDP
Catalog No. 50973185
Encompass_Preferred
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has five necks: (1) 24/40 center, (4) 24/40 sides.
Catalog No. 50-973-185 Supplier Ace Glass, Inc. Supplier No. 652923
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has five necks: (1) 24/40 center, (4) 24/40 sides.