Learn More
Ace Glass, Inc. Head, 6in, five neck, 29/42 center, (4)29/42
SDP
Catalog No. 50973187
Encompass_Preferred
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has five necks: (1) 29/42 center, (4) 29/42 sides.
Catalog No. 50-973-187 Supplier Ace Glass, Inc. Supplier No. 652927
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has five necks: (1) 29/42 center, (4) 29/42 sides.