Learn More
Ace Glass, Inc. Head, 6in, four neck, 29/42 center, (2)24/40, 10/30
SDP
Catalog No. 50973183
Encompass_Preferred
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has four necks: (1) 29/42 center, (2) 24.40 sides, (1) 10/30 side.
Catalog No. 50-973-183 Supplier Ace Glass, Inc. Supplier No. 652904
6 inch reaction head with a flat ground flange for use with all reaction flasks that have a 150mm (6 inch) Duran ground mating flange. Head has four necks: (1) 29/42 center, (2) 24.40 sides, (1) 10/30 side.