Learn More
Ace Glass, Inc. Head, 24/40 center, (2)24/40 sides, (1)10/30, 137mm OD x 100mm ID flat flange
SDP
Catalog No. 50973130
Encompass_Preferred
4 neck, flat-flange head for use with 1-4L cylindrical reaction flasks. Supplied with 24/40 reinforced outer center neck, and (2) 24/40 reinforced outer joints in-line, and a 10/30 reinforced outer side joint for thermometer. 137mm O.D. x 100mm I.D. flat flange.
Catalog No. 50-973-130 Supplier Ace Glass, Inc. Supplier No. 651546
4 neck, flat-flange head for use with 1-4L cylindrical reaction flasks. Supplied with 24/40 reinforced outer center neck, and (2) 24/40 reinforced outer joints in-line, and a 10/30 reinforced outer side joint for thermometer. 137mm O.D. x 100mm I.D. flat flange.