Learn More
BURKLE INC Head, volume 0.01 oz. (0,2 ml), MicroSampler tube-Ø 0.47 in. (12 mm)
Catalog No. 5000025908
Encompass_Preferred
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
Catalog No. 50-000-25908 Supplier BURKLE INC Supplier No. 53070102
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.