Learn More
BURKLE INC Novartos V4A, head
Catalog No. 5000026056
Encompass_Preferred
The non-rounded tip for Novartos can be replaced on the sampling lance. The very pointed and stable tip can be used to pierce directly into bags or other containers.
Catalog No. 50-000-26056 Supplier BURKLE INC Supplier No. 53581011
The non-rounded tip for Novartos can be replaced on the sampling lance. The very pointed and stable tip can be used to pierce directly into bags or other containers.