Learn More
BURKLE INC Head, volume 0.02 oz. (0,5 ml), MicroSampler tube-Ø 0.47 in. (12 mm)
Catalog No. 5000025761
Encompass_Preferred
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.
Catalog No. 50-000-25761 Supplier BURKLE INC Supplier No. 53070105
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.