Learn More
Ace Glass, Inc. Head, 60mm Duran flange, 5 neck, 24/40 center, (3)14/20, (1)24/40
SDP
Catalog No. 50972985
Encompass_Preferred
5 neck head with 60mm flat ground Duran flange. Head features one 24/40 center neck, three 14/20 side necks, and one 24/40 side neck.
Catalog No. 50-972-985 Supplier Ace Glass, Inc. Supplier No. 644302
5 neck head with 60mm flat ground Duran flange. Head features one 24/40 center neck, three 14/20 side necks, and one 24/40 side neck.