Learn More
Ace Glass, Inc. Head, 60mm Duran flange, 5 neck, 29/42 center, (3)14/23, (1)24/29
SDP
Catalog No. 50972991
Encompass_Preferred
5 neck head with 60mm flat ground Duran flange. Head features one 29/42 center neck, three 14/23 side necks, and one 24/29 side neck.
Catalog No. 50-972-991 Supplier Ace Glass, Inc. Supplier No. 6443106
5 neck head with 60mm flat ground Duran flange. Head features one 29/42 center neck, three 14/23 side necks, and one 24/29 side neck.