Learn More
Ace Glass, Inc. Head, 60mm Duran flange, 5 neck, 29/42 center, (1)24/40, (3)14/20
SDP
Supplier:  Ace Glass, Inc. 644304
Encompass_Preferred
5 neck head with 60mm flat ground Duran flange. Head features one 29/42 center neck, three 14/20 side necks, and one 24/40 side neck.
Catalog No. 50-972-986