Learn More
Thermo Scientific Thermal Head Assembly Primary Series Pack
Catalog No. NC0551209 Shop All Thermo Scientific Products
Encompass
Head assembly for mass spectrometer contains ion source, mass analyzer, and detector enabling instrument to ionize, separate, detect gas phase ions by mass; modular component allows changing head on instrument frame for maintenance or new capabilities, impacting performance parameters like resolution, accuracy, sensitivity, range; attaches to frame through mounting interfaces and makes electrical connections.
This item is not returnable. View return policy
Catalog No. NC0551209 Supplier Thermo Scientific Supplier No. 057937
Head assembly for mass spectrometer contains ion source, mass analyzer, and detector enabling instrument to ionize, separate, detect gas phase ions by mass; modular component allows changing head on instrument frame for maintenance or new capabilities, impacting performance parameters like resolution, accuracy, sensitivity, range; attaches to frame through mounting interfaces and makes electrical connections.