Learn More
Ace Glass, Inc. Head, distribution, model R220, glassware sets C, RB and R, part 41373
SDP
Catalog No. 50978238
Encompass
Borosilicate glass, lower distribution head for model R220 rotary evaporator. For use with glassware sets C, RB, and R, part 41373. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included.
Catalog No. 50-978-238 Supplier Ace Glass, Inc. Supplier No. 397005
Borosilicate glass, lower distribution head for model R220 rotary evaporator. For use with glassware sets C, RB, and R, part 41373. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included.