Learn More
Ace Glass, Inc. Head, distribution, models R220EX&SE, glassware sets C, RB and R, part 46511
SDP
Catalog No. 50971496
Encompass
Borosilicate glass, lower distribution head for rotary evaporator models R220EX & R220SE. For use with glassware sets C, RB, and R, part 46511. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included
Catalog No. 50-971-496 Supplier Ace Glass, Inc. Supplier No. 397010
Borosilicate glass, lower distribution head for rotary evaporator models R220EX & R220SE. For use with glassware sets C, RB, and R, part 46511. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included