Learn More
Ace Glass, Inc. Head, distribution, models R220EX&SE, glassware sets D, D2, DB and DB2, part 41307
SDP
Catalog No. 50971498
Encompass
Borosilicate glass, lower distribution head for rotary evaporator models R220EX & R220SE. For use with glassware sets D, D2, DB, and DB@, part 41307. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included.
Catalog No. 50-971-498 Supplier Ace Glass, Inc. Supplier No. 397018
Borosilicate glass, lower distribution head for rotary evaporator models R220EX & R220SE. For use with glassware sets D, D2, DB, and DB@, part 41307. Head has an improved design by utilizing Ace-Threds for connections. Inner ball joint has a CAPFE o-ring. All caps are included.