Learn More
International Enviroguard™ Head and Face Hoods (Ninja Hoods)
Catalog No. 19472782 Shop All International Enviroguard Systems Products
Catalog No. 19-472-782 Supplier International Enviroguard™ Supplier No. NH2000
Add to Cart
Add to Cart
These Head and Face Hoods (Ninja Hoods) provide economical, lightweight protection against dirt, grime and particulates in non-hazardous environments.
These head and face covers (''Ninja Hoods'') offer dual protection - they protect your face from particulates while also protecting workplace products from contaminants you may generate such as saliva, hair, dander, and more. Wearing a head and face hood is a practical, win-win for you and your workplace. Better yet, this one product can replace separate items such as beard restraints and hair bouffant.
  • Lightweight polypropylene product that is an excellent one-piece replacement where separate beard restraints and hair bouffants are used
  • Common applications include food processing, food service, pharmaceuticals, cleanrooms and controlled environments, agriculture, healthcare, and more