Learn More
Ace Glass, Inc. Head, 100mm Duran flange, O-ring groove, #15 center, (3)#15, (1)#7
SDP
Catalog No. NC0634289
Encompass_Preferred
100mm head with Schott-style flange, for pressure or filter reactors. It also fits 1000-2000mL flasks. Center neck is #15 Ace-Thred, three vertical #15 Ace-Thred side necks, plus one angled #7 Ace-Thred side neck. Pressure: 35 psi.
Catalog No. NC0634289 Supplier Ace Glass, Inc. Supplier No. 643335
100mm head with Schott-style flange, for pressure or filter reactors. It also fits 1000-2000mL flasks. Center neck is #15 Ace-Thred, three vertical #15 Ace-Thred side necks, plus one angled #7 Ace-Thred side neck. Pressure: 35 psi.