Learn More
Discovery Life Sciences Head Neck Ca Active Tx PBMC
SDP
Catalog No. 5000804075
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04075 Supplier Discovery Life Sciences Supplier No. 120650
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays