Learn More
Discovery Life Sciences Head Neck Cancer PD L1 DTC
Supplier: Discovery Life Sciences 321210.2
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics