Learn More
Ace Glass, Inc. HEAD ONLY HEAD ONLY
SDP
Catalog No. 50976754
Encompass
Head only with 14/20 joint, used with distillation head with finger type condenser.
Catalog No. 50-976-754 Supplier Ace Glass, Inc. Supplier No. 935802
Head only with 14/20 joint, used with distillation head with finger type condenser.