Learn More
Bellco Glass, Inc. Head Plate Setup Stand Bioreactor
SDP
Catalog No. 501944767
Encompass_Preferred
Headplate setup stand is for hands-free assembly of bioreactor head plates manufactured from aluminum and measures 8"x8"x3" assembled. Headplate is inserted into the slot on the stand and secured into place with thumbscrews. It holds any head plate up to 6mm thick. MADE IN THE USA
Catalog No. 50-194-4767 Supplier Bellco Glass, Inc. Supplier No. 196410000
Headplate setup stand is for hands-free assembly of bioreactor head plates manufactured from aluminum and measures 8"x8"x3" assembled. Headplate is inserted into the slot on the stand and secured into place with thumbscrews. It holds any head plate up to 6mm thick. MADE IN THE USA