Learn More
Chemglass Life Sciences Head-Plate Assembly Stand ONLY, for Bioreactors
SDP
Catalog No. 501294460
Encompass
Constructed of blue anodized aluminum, stand permits easy assembly of components onto head-plates.Will fit from 2 to 15 liter head-plates, which are secured with 4 nylon tipped thumb screws.Plate is 8" wide x 8" deep.Distance between uprights is 8mm.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4460 Supplier Chemglass Life Sciences Supplier No. CLS1380100
Constructed of blue anodized aluminum, stand permits easy assembly of components onto head-plates.Will fit from 2 to 15 liter head-plates, which are secured with 4 nylon tipped thumb screws.Plate is 8" wide x 8" deep.Distance between uprights is 8mm.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.