Learn More
Ace Glass, Inc. Head, PTFE, 100mm, 7 neck, 3/4in center, (1) 1in 10° angled, (2)3/4in vertical, (3) 1/4in vertical
SDP
Supplier:  Ace Glass, Inc. 1286025
Encompass
100mm PTFE heads allow for various combinations of jointed and/or compression fitting openings as well as the ability to plug unused openings. For use with any Duran-style, o-ring grooved flanged reaction vessels
Catalog No. 50-977-885