Learn More
Ace Glass, Inc. 150mm PTFE head, (1) 1" center neck, (3) 1" vertical necks, (1) 1" 10 x 10° compound angle neck, (4) 1/2" vertical necks
SDP
Catalog No. 50983467
Encompass
150mm PTFE head allows for various combinations of jointed and/or compression fittings, as well as, the ability to plug unused openings. PTFE head has 9 openings: (1) vertical 1 center neck, (3) vertical 1 side necks, (1) 10 x 10 angled 1 side neck, and (4) 1/2 vertical side necks. Head can be used with any Duran-style, o-ring groove flange reaction vessels.
Catalog No. 50-983-467 Supplier Ace Glass, Inc. Supplier No. 1286216
150mm PTFE head allows for various combinations of jointed and/or compression fittings, as well as, the ability to plug unused openings. PTFE head has 9 openings: (1) vertical 1 center neck, (3) vertical 1 side necks, (1) 10 x 10 angled 1 side neck, and (4) 1/2 vertical side necks. Head can be used with any Duran-style, o-ring groove flange reaction vessels.