Learn More
Sigma Aldrich Fine Chemicals Biosciences Head set for Vortex-GenieR
Catalog No. 502904976
Encompass
for continuous vortexing of beakers flasks and tubes. Includes a recessed platform elastic flask harness and an assortment of tube inserts (accepts 10 to 37mm diam. tubes).
Catalog No. 50-290-4976 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2584581PAK
for continuous vortexing of beakers flasks and tubes. Includes a recessed platform elastic flask harness and an assortment of tube inserts (accepts 10 to 37mm diam. tubes).