Learn More
Perkin Elmer US LLC 20 mL Clear Glass Crimp Top Vial with Write-On Patch, 1000/pk
Catalog No. NC0622629
Encompass_Preferred
PerkinElmer Headspace vials are manufactured to specific tolerances that are guaranteed to fit and perform with PerkinElmer and non-PerkinElmer instruments.
Catalog No. NC0622629 Supplier Perkin Elmer US LLC Supplier No. N9303348
PerkinElmer Headspace vials are manufactured to specific tolerances that are guaranteed to fit and perform with PerkinElmer and non-PerkinElmer instruments.