Learn More
Electron Microscopy Sciences Headband Magnifier, 2.50x, 8" with auxillary 3", 8" with auxillary 3" range
SDP
Supplier:  Electron Microscopy Sciences 6810806
Encompass_Preferred
Lightweight, adjustable headband for comfort provides close-up magnification while leaving both hands free for working. Tilt up and away when not in use. Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue. Includes 3.5X Magnification Auxiliary Lens.
Magnification: 2.50x
Range: 8", auxillary 3"
Catalog No. 50-264-60