Learn More
Fine Science Tools Headband Magnifiers, 2.75x, 15 cm Working Distance
SDP
Catalog No. NC9385811
Encompass
Product: Headband Magnifiers/ Weight: 140g/ Magnification: 2.75x/ Working Distance: 15cm/ Unit: Each
Catalog No. NC9385811 Supplier Fine Science Tools Supplier No. 2803003
Product: Headband Magnifiers/ Weight: 140g/ Magnification: 2.75x/ Working Distance: 15cm/ Unit: Each