Learn More
MSC HEADCOVER PAPR KIT
Catalog No. 5000685579
Encompass
The 3M Versaflo Healthcare PAPR Kit TR-600-HKL provides versatile respiratory protection against gases, vapors, and particulates, specifically designed for healthcare and pharmaceutical environments.
  • Headgear type: headcover
  • System type: PAPR
  • Application: healthcare, pharmaceutical
  • Headgear material: polypropylene
  • Adjustment: adjustable
  • Size: medium
  • Breathing tube type: BT-30
  • PAPR assembly type: TR-602N
  • Includes: S-133S headcover, single station battery charger kit, easy clean belt, storage and cleaning kit, filter cover
Catalog No. 50-006-85579 Supplier MSC Supplier No. 10047108
The 3M Versaflo Healthcare PAPR Kit TR-600-HKL provides versatile respiratory protection against gases, vapors, and particulates, specifically designed for healthcare and pharmaceutical environments.
  • Headgear type: headcover
  • System type: PAPR
  • Application: healthcare, pharmaceutical
  • Headgear material: polypropylene
  • Adjustment: adjustable
  • Size: medium
  • Breathing tube type: BT-30
  • PAPR assembly type: TR-602N
  • Includes: S-133S headcover, single station battery charger kit, easy clean belt, storage and cleaning kit, filter cover