Learn More
MSC North PAPR Hood, Primair Head Cover, Medium, Tyvek, 1 ea
Catalog No. 502370347
Encompass
This PAPR hood includes a Primair loose fitting head cover designed for respiratory protection in various work environments.
  • Headgear type: Hood
  • Headgear material: Tyvek
  • System compatibility: PAPR
  • Size: Medium
  • Series compatibility: CA101; CA101D
  • Color: Yellow
  • Contents: Primair loose fitting head cover
Catalog No. 50-237-0347 Supplier MSC Supplier No. 63943146
This PAPR hood includes a Primair loose fitting head cover designed for respiratory protection in various work environments.
  • Headgear type: Hood
  • Headgear material: Tyvek
  • System compatibility: PAPR
  • Size: Medium
  • Series compatibility: CA101; CA101D
  • Color: Yellow
  • Contents: Primair loose fitting head cover