Learn More
Med Vet International Wraparound Face Shield 3M One Size Fits Most Full Length Impact Resistant Reusable Nonsterile, 82782-00000, 5/Case
Catalog No. 502831314
Encompass_Preferred
A complete headgear and faceshield safety system, this high-strength thermoplastic hard hat helps provide durable impact resistance and reliable head and face protection over a wide range of temperatures Adjustable ratchet suspension Safety system comes complete with both headgear and faceshield Meets the requirements of ANSI Z87Five position crown strap and thirteen position ratchet suspension offer a customized fit for individual wearers Offers head and face protection for many jobs and tasks where liquid and chemical splashes may occur
Catalog No. 50-283-1314 Supplier Med Vet International Supplier No. 1063707CS
A complete headgear and faceshield safety system, this high-strength thermoplastic hard hat helps provide durable impact resistance and reliable head and face protection over a wide range of temperatures Adjustable ratchet suspension Safety system comes complete with both headgear and faceshield Meets the requirements of ANSI Z87Five position crown strap and thirteen position ratchet suspension offer a customized fit for individual wearers Offers head and face protection for many jobs and tasks where liquid and chemical splashes may occur