Learn More
Perkin Elmer US LLC Headspace Mini Starter Kit
Catalog No. 502103945
Encompass_Preferred
Includes the following: (1) 20 mm Hand Crimper (100) 20 mL Clear Glass Crimp Top Vials (100) PTFE/Butyl (red) Septa with Pre-Assembled Aluminum Crimp Caps (100) PTFE/Silicone (white) Septa with Pre-Assembled Aluminum Crimp Caps (100) Aluminum/Silicone Septa with Pre-Assembled Aluminum Crimp Caps (100) 20 mL Clear Glass Screw Top Vials (100) PTFE/Butyl (red) Septa with Steel Magnetic Screw Caps (100) PTFE/Silicone (white) Septa with Steel Magnetic Screw Caps (2) Needle Seal Assemblies (10) O-Rings (1) Pressure Gaue with Needle for Vials
Catalog No. 50-210-3945 Supplier Perkin Elmer US LLC Supplier No. N6710197
Includes the following: (1) 20 mm Hand Crimper (100) 20 mL Clear Glass Crimp Top Vials (100) PTFE/Butyl (red) Septa with Pre-Assembled Aluminum Crimp Caps (100) PTFE/Silicone (white) Septa with Pre-Assembled Aluminum Crimp Caps (100) Aluminum/Silicone Septa with Pre-Assembled Aluminum Crimp Caps (100) 20 mL Clear Glass Screw Top Vials (100) PTFE/Butyl (red) Septa with Steel Magnetic Screw Caps (100) PTFE/Silicone (white) Septa with Steel Magnetic Screw Caps (2) Needle Seal Assemblies (10) O-Rings (1) Pressure Gaue with Needle for Vials