Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 18 mm Screw, Round Bottom, No Patch

Supplier:  Phenomenex Inc AR5328012

Encompass_Preferred

Headspace Vial, 23x75mm, 20mL, Clear 33 AR5-3280-12

Catalog No. 50-239-8944


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.