Learn More
BTNX Inc Healgen COVID-19/Flu A&B Ag Combo Rapid Test Cassette (Swab) 25 Tests/Kit
Catalog No. NC3957389
Encompass_Preferred
The Healgen COVID-19/Flu A&B Ag Combo Rapid Test Cassette (swab) is a lateral flow immunochromatographic assay intended for in vitro rapid, simultaneous qualitative detection and differentiation of influenza A and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigen directly from anterior nasal swab specimens. CLIA-Waived. Time to result: 15 minutes. Format: Cassette. Quantity: 25/kit.The Healgen COVID-19/Flu A&B Ag Combo Rapid Test Cassette (swab) is intended for use in individuals who are suspected of respiratory viral infection consistent with COVID-19 by their healthcare provider within the first five (5) days of symptom onset when tested at least twice over three days with at least 48 hours between tests.Suitable for testing individuals 2+ years old. Swabbing should be performed by an adult for children aged 2 to 13. User must be aged 14 + to perform self test.
Catalog No. NC3957389 Supplier BTNX Inc Supplier No. COF19CPU25G
The Healgen COVID-19/Flu A&B Ag Combo Rapid Test Cassette (swab) is a lateral flow immunochromatographic assay intended for in vitro rapid, simultaneous qualitative detection and differentiation of influenza A and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigen directly from anterior nasal swab specimens. CLIA-Waived. Time to result: 15 minutes. Format: Cassette. Quantity: 25/kit.The Healgen COVID-19/Flu A&B Ag Combo Rapid Test Cassette (swab) is intended for use in individuals who are suspected of respiratory viral infection consistent with COVID-19 by their healthcare provider within the first five (5) days of symptom onset when tested at least twice over three days with at least 48 hours between tests.Suitable for testing individuals 2+ years old. Swabbing should be performed by an adult for children aged 2 to 13. User must be aged 14 + to perform self test.