Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Assure Tech Co., Ltd. ASSURE COVID-19 IgG/IgM Rapid Test Device

Catalog No. 23111397
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-397 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-397 Supplier Assure Tech Co., Ltd. Supplier No. COVW23

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

20 tests/kit, disposable pipettes, sterile safety lancets, alcohol prep pads, buffer. Note: this test requires a control (not included).

Provides results in 15 minutes with 20 tests/kit.

  • Highly accurate, easy to use rapid test kit provides results in 15 minutes
  • Includes:
    • 20 tests/kit
    • Disposable pipettes
    • Sterile safety lancets
    • Alcohol prep pads
    • Buffer
    • Package Insert
  • Intended use:
    • Rapid lateral flow chromatographic immunoassay for qualitative detection and differentiation of IgM and IgG antibodies to SARS-CoV-2 in human venous whole blood (sodium EDTA), serum, plasma (sodium EDTA) and fingerstick whole blood
    • Identify individuals with an adaptive immune response to SARS-CoV-2
  • Limited use:
    • Laboratories certified under the Clinical Laboratory Improvement Amendments of 1988 (CLIA), 42 U.S.C. 263a
  • Note:
    • Device should not be used to diagnose acute SARS-CoV-2 infection
    • FDA EUA CLIA Waived authorization- fingerstick blood samples can now be tested in POC (Point of Care) setting such as doctor's offices, hospitals, urgent care centers and emergency rooms rather than having to be sent to a central lab for testing. under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation

Specifications

Content And Storage Ambient
Control Sets This test requires a control (not included).
CE Marker Yes
Clia Complexity Waived (FDA, EUA)
Detectable Analytes SARS-CoV-2 IgG, IgM
Sample Type Human venous whole blood (sodium EDTA), serum, plasma (sodium EDTA) and fingerstick whole blood
Sensitivity 100% Combined Sensitivity
Type Covid-19 Antibody
Disposable Single-use
Includes 20 tests/kit, disposable pipettes, sterile safety lancets, alcohol prep pads, buffer, and a Pk. insert
Quantity 20 Tests/Kit
Specificity Combined specificity 98.8%
Test Time 15 min.
Show More Show Less

FDA EUA CLIA WAIVED

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.