Learn More
3M Health Care Particulate Respirator and Surgical Mask
Catalog No. 22355007
Change view
Click to view available options
Fastening Type:
Headband
Catalog No. Fastening Type
22-355-007 Headband
1 options
This item is not returnable. View return policy
Catalog No. 22-355-007 Supplier 3M Supplier No. MMM 1860
Add to Cart
Add to Cart
This item is not returnable. View return policy
This health care particulate respirator and surgical mask helps provide respiratory protection against certain airborne biological particles. It is disposable and fluid resistant to splash and spatter of blood and other infectious material.
  • NIOSH Approved: N95
  • FDA Cleared for use as surgical mask
  • Helps protect against certain airborne biological particles
  • Fluid resistant and disposable
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Headband
Certifications/Compliance NIOSH 42 CFR 84, N95, Approval Number: TC-84 A-0006
Disposable Single-use
Model 1860
Resistance Fluid Resistant