Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

binx™ health io™ Vaginal Swab Specimen Collection Kit

Catalog No. 22021833
Change view
Click to view available options
Clia Complexity:
Waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-021-833 Waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-833 Supplier binx™ Supplier No. 5.012.001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Vaginal Swab Specimen Collection Kit is required for the collection, transportation, and preservation of DNA in a vaginal swab sample for use with the binx™ health io™ CT/NG Assay.

The binx™ health io™ Vaginal Swab Specimen Collection is required for the collection, transportation, and preservation of DNA in a vaginal swab specimen for use with the binx™ health io™ CT/NG Assay.

  • Kit consists of a screw cap tube containing Transport and Preservation Medium and a sample collection swab
  • Specimens stored and transported using the binx™ health io™ Vaginal Swab Specimen Collection Kit are intended to be processed using the binx™ health io™ CT/NG Assay using the binx™ health io™ Instrument
  • It is not intended to be used for the preservation and transfer of samples for any other test
  • Patient samples may be stored in the Collection Kit for 24 hours at room temperature or for seven (7) days refrigerated at 2°C-8°C

Specifications

Clia Complexity Waived
Detectable Analytes C. trachomatis, N. gonorrhoeae
Sample Type Vaginal Swab
Sensitivity High
Type Collection Kit
For Use With (Application) Chlamydia Testing
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.