Learn More
Enterprise Technology Solutions Health-O-Meter Digital Scale, w/Remote Display, 14"x17-1/4"x2", Black/Gray
SDP
Catalog No. 5000828287
Encompass_Preferred
Digital scale with remote digital display provides professional-grade accuracy in a small footprint. The platform can be placed virtually anywhere. The corded display unit can be set on a countertop or mounted on a wall with the included hardware. Advanced features such as EMR connectivity via USB, hold and tare will allow you quickly take and record weight readings from the 1" LCD screen. Other functions include Body Mass Index (BMI), auto off, auto zero, Lb/Kg conversion, zero out, and LB/KG Lock-Out. The Digital Physicians Scale also offers a weight capacity of 500 lb. (220 kg) and 17-3/4" wide x 14" deep platform. Digital scale runs on six AA batteries (included) or a 120V Adapter (not included).
Catalog No. 50-008-28287 Supplier Enterprise Technology Solutions Supplier No. HHM498KL
Digital scale with remote digital display provides professional-grade accuracy in a small footprint. The platform can be placed virtually anywhere. The corded display unit can be set on a countertop or mounted on a wall with the included hardware. Advanced features such as EMR connectivity via USB, hold and tare will allow you quickly take and record weight readings from the 1" LCD screen. Other functions include Body Mass Index (BMI), auto off, auto zero, Lb/Kg conversion, zero out, and LB/KG Lock-Out. The Digital Physicians Scale also offers a weight capacity of 500 lb. (220 kg) and 17-3/4" wide x 14" deep platform. Digital scale runs on six AA batteries (included) or a 120V Adapter (not included).