Learn More
Enterprise Technology Solutions Health-O-Meter Physician Digital Scale, 1" LCD, 14-1/8"x21"x53-5/8", Silver
SDP
Catalog No. 5000828283
Encompass_Preferred
Eye-level digital scale features Electronic Medical Record (EMR) connectivity via USB to interface with a computer, monitor, printer or other electronic device for fast, easy and accurate data transfer. Capacity is 500 lb. with a 0.2 lb. resolution. 1" LCD is easy to read. Functions include LB/KG conversion, LB/KG lockout, automatic Body Mass Index (BMI) calculation, Zero Out/Tare, Hold/Release, Auto Zero and Auto Off. Platform size is 12-3/4" wide x 15-1/2" deep. Easy-to-use, built-in height rod measures 30" to 84-1/4" (76cm to 214cm). Digital scale features two wheels for easy relocation and includes six AA batteries.
Catalog No. 50-008-28283 Supplier Enterprise Technology Solutions Supplier No. HHM500KL
Eye-level digital scale features Electronic Medical Record (EMR) connectivity via USB to interface with a computer, monitor, printer or other electronic device for fast, easy and accurate data transfer. Capacity is 500 lb. with a 0.2 lb. resolution. 1" LCD is easy to read. Functions include LB/KG conversion, LB/KG lockout, automatic Body Mass Index (BMI) calculation, Zero Out/Tare, Hold/Release, Auto Zero and Auto Off. Platform size is 12-3/4" wide x 15-1/2" deep. Easy-to-use, built-in height rod measures 30" to 84-1/4" (76cm to 214cm). Digital scale features two wheels for easy relocation and includes six AA batteries.