Learn More
Enterprise Technology Solutions Health-O-Meter Scale, Digital, Glass, 13"Wx13"Lx7/8"H, Black
SDP
Catalog No. 5000828274
Encompass_Preferred
The Digital Glass Scale features an anti-slip surface for extra traction and peace of mind. High weight capacity reads up to 440 lb. for more versatile use. Five-second weight hold displays the weight for five seconds after stepping off to give the user or caregiver time to stabilize themselves or the patient. Backlit display makes it easy to read the weight from any angle and in any light. Medical-grade scale is designed for use in the home or bathroom with accuracy and benefits of a professional scale.
Catalog No. 50-008-28274 Supplier Enterprise Technology Solutions Supplier No. HHM810KL
The Digital Glass Scale features an anti-slip surface for extra traction and peace of mind. High weight capacity reads up to 440 lb. for more versatile use. Five-second weight hold displays the weight for five seconds after stepping off to give the user or caregiver time to stabilize themselves or the patient. Backlit display makes it easy to read the weight from any angle and in any light. Medical-grade scale is designed for use in the home or bathroom with accuracy and benefits of a professional scale.