Learn More
Enterprise Technology Solutions Health-O-Meter Scale, w/ Height Rod, 20-1/4"x10-3/8"x57-1/2", Black/Silver
SDP
Catalog No. 5000828272
Encompass_Preferred
Scale with height rod features easy-to-read, rotating poise bars that offer duel reading in engraved pounds and kilograms. Engraved numerals on the poise bars can withstand frequent use, providing long-lasting readability. Imperial graduations are engraved by 1/4 lb. Scale capacity is 390 lb. or 180 kg. Easy-to-use, height rod measures from 23-5/8" to 84" (60cm to 213cm). Scale is factory calibrated. Platform measures 10-1/2" wide x 14" deep and has a black nonslip mat. Scale is made of powder-coated steel to prevent rust inside and out for extended life.
Catalog No. 50-008-28272 Supplier Enterprise Technology Solutions Supplier No. HHM402KL
Scale with height rod features easy-to-read, rotating poise bars that offer duel reading in engraved pounds and kilograms. Engraved numerals on the poise bars can withstand frequent use, providing long-lasting readability. Imperial graduations are engraved by 1/4 lb. Scale capacity is 390 lb. or 180 kg. Easy-to-use, height rod measures from 23-5/8" to 84" (60cm to 213cm). Scale is factory calibrated. Platform measures 10-1/2" wide x 14" deep and has a black nonslip mat. Scale is made of powder-coated steel to prevent rust inside and out for extended life.