Learn More
Description
- 1200 mL refill
- Exceptionally mild foam hand soap designed for frequent hand washing.
- Helps wash away dirt and germs commonly found in a healthcare environment
- Designed to maintain skin health with frequent use in a healthcare environment
- ECOLOGO™ Certified to Standard Name UL 2784
- USDA Certified Biobased Product
- Enriched with a blend of moisturizers and skin conditioners
- With natural moisturizers
- Hypoallergenic
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Blue |
| Form | Foam |
| For Use With (Equipment) | PURELL ES8 Touch-Free Soap Dispensers |
| Size | 1200 mL |
| Volume (Metric) | 1200 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

